missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO2 Antibody (OTI1A4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72453B
This item is not returnable.
View return policy
Description
IDO2 Monoclonal specifically detects IDO2 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| IDO2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| IDO2 | |
| Full length recombinant protein of human IDO2 (NP_919270) produced in HEK293T cells. | |
| 0.1 mL | |
| 169355.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI1A4 | |
| Western Blot, Flow Cytometry | |
| EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction