Learn More
IDO2 Mouse anti-Human, Clone: OTI12G4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806163
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Along with the enzymes encoded by the INDO and TDO2 genes, the enzyme encoded by the INDOL1 gene metabolizes tryptophan in the kynurenine pathway.Specifications
| IDO2 | |
| Monoclonal | |
| Unconjugated | |
| IDO2 | |
| INDOL1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 169355 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI12G4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q6ZQW0 | |
| IDO2 | |
| Full length human recombit protein of human IDO2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.