Learn More
PKLR Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97071F
Product Code. 18086166
  • € 415.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
PKLR Polyclonal specifically detects PKLR in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
PKLR
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase
Produced in rabbits immunized with purified, recombinant PKLR (NP_000289.1; Leu47-Ser574).
0.1 mL
Chaperone Mediated Autophagy (CMA), Lipid and Metabolism, Signal Transduction
5313
Store at 4°C in the dark.
IgG
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC