missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IdU Mouse anti-Chemical, Clone: OTI6E4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA190199
This item is not returnable.
View return policy
Description
Idu (Idoxuridine) is a nucleoside analogue, a modified form of deoxyuridine, similar enough to be incorporated into viral DNA replication, but the iodine atom added to the uracil component blocks base pairing. It is used only topically due to cardiotoxicity. It was synthesized by William Prusoff in the late 1950s.Specifications
| IdU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI6E4 | |
| Unconjugated | |
| IdU | |
| Iododeoxyuridine coupled to keyhole limpet hemocyanin. | |
| 100 μL | |
| Primary | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction