missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2-Sulfatase/IDS Antibody (OTI1D9) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70994
This item is not returnable.
View return policy
Description
Iduronate 2-Sulfatase/IDS Monoclonal specifically detects Iduronate 2-Sulfatase/IDS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| Iduronate 2-Sulfatase/IDS | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
| Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS | |
| Mouse | |
| 59.2 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| OTI1D9 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| IDS | |
| Full length human recombinant protein of human IDS(NP_000193) produced in HEK293T cell. | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 3423 | |
| Human, Mouse, Rat, Monkey | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction