Learn More
Thermo Scientific™ IEF Cathode buffer (10X) with lysine and arginine
€ 42.15 - € 92.80
Specifications
Form Liquid
Storage Requirements Ambient temperatures
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440237
SDS View Documents Promotion Details
Thermo Scientific™
J62204.AP
500mL
€ 92.80
500mL
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
15430237
SDS View Documents Promotion Details
Thermo Scientific™
J62204.AK
250mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Liquid
Ambient temperatures