missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IER3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38513
This item is not returnable.
View return policy
Description
IER3 Polyclonal specifically detects IER3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IER3 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P46695 | |
| IER3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MCHSRSCHPTMTILQAPTPAPSTIPGPRRGSGPEIFTFDPL | |
| 0.1 mL | |
| Apoptosis | |
| 8870 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DIF-2, DIF2GLY96, Differentiation-dependent gene 2 protein, expressed in pancreatic carcinoma, gly96, mouse, homolog of, IEX-1, IEX1death protein, IEX-1L, Immediate early protein GLY96, immediate early response 3, Immediate early response 3 protein, immediately early gene X-1, PRG1PACAP-responsive gene 1 protein, Protein DIF-2, Protein PRG1, radiation-inducible immediate-early gene IEX-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction