missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IER3IP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15847863
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847863 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15847863 Supplier Invitrogen™ Supplier No. PA569453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a small protein that is localized to the endoplasmic reticulum (ER) and may play a role in the ER stress response by mediating cell differentiation and apoptosis. Transcription of this gene is regulated by tumor necrosis factor alpha and specificity protein 1 (Sp1). Mutations in this gene may play a role in microcephaly, epilepsy, and diabetes syndrome (MEDS), and a pseudogene of this gene is located on the long arm of chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IER3IP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IER3IP1
Gene Accession No. Q9CR20
Gene Alias 1110057H19Rik; 796; AI644142; AL022842; AL022863; AV026606; HSPC039; IER3IP1; immediate early response 3 interacting protein 1; immediate early response 3-interacting protein 1; MEDS; PRO2309; uncharacterized protein LOC500270
Gene Symbols IER3IP1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse IER3IP1 (aa 9-58).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66191
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.