Learn More
Novus Biologicals™ IF3EI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30051
Product Code. 30045920
  • € 571.00 / 100µg
Description
IF3EI Polyclonal antibody specifically detects IF3EI in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IF3EI
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Immunoprecipitation, Western Blot
Unconjugated
eIEF associated protein HSPC021, EIF3EIP, eIF3l, EIF3S11, EIF3S6IP, Eukaryotic translation initiation factor 3 subunit 6-interacting protein, Eukaryotic translation initiation factor 3 subunit E-interacting protein, eukaryotic translation initiation factor 3 subunit L, eukaryotic translation initiation factor 3, subunit 6 interacting protein, eukaryotic translation initiation factor 3, subunit L, HSPC021, HSPC025, MSTP005, subunit E interacting protein
Between 514 and 564
100 μg
Core ESC Like Genes, Stem Cell Markers
51386
Store at 4°C. Do not freeze.