missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFI6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16604075
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16604075 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16604075 Supplier Invitrogen™ Supplier No. PA599824

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFI6.

G1P3, also known as Interferon alpha-inducible protein 6 (IFI6) and Interferon-induced protein 6-16, is a 130 amino acid member of the IFI6 family of proteins. Localized to the mitochondria, G1P3 is a multi-pass membrane protein that is induced by IFN-α2β. G1P3 has been shown to play a major role in the apoptosis pathway. Specifically, G1P3 acts as a cell survival protein by inhibiting caspase-3 activity, which antagonizes apoptosis. G1P3 has been implicated in tumorigenesis and is the subject of therapeutic studies, as reducing G1P3-mediated antiapoptotic signals could suggest improved therapies for myeloma or other malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFI6
Gene Accession No. P09912
Gene Alias 6-16; FAM14C; G1P3; IFI6; IFI616; IFI-6-16; interferon alpha inducible protein 6; interferon alpha-inducible protein 6; interferon, alpha-inducible protein 6; interferon, alpha-inducible protein clone IFI-6-16; interferon-induced protein 6-16
Gene Symbols IFI6
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFI6(Accession P09912), corresponding to amino acid residues A5-G55.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2537
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.