missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIH1 Monoclonal Antibody (OTI12A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803671
This item is not returnable.
View return policy
Description
Melanoma differentiation associated gene 5, MDA5, is a pattern recognition receptor to transmit the signal.Once activated, both proteins signal through IPS-1 activating transcription factors NF- kappaB and IRF-3 and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus, paromyxoviruses, Japanese encephalitis virus. MDA5 is essential for signaling by picornavirus. Both RIG-I and MDA5 are responsible for signaling by West Nile virus.Specifications
| IFIH1 | |
| Monoclonal | |
| Unconjugated | |
| IFIH1 | |
| AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64135 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI12A6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BYX4 | |
| IFIH1 | |
| Full length human recombit protein of human IFIH1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction