missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIH1 Monoclonal Antibody (OTI12A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803671S
This item is not returnable.
View return policy
Description
IFIH1 Monoclonal antibody specifically detects IFIH1 in Human samples. It is validated for Western BlotSpecifications
| IFIH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BYX4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64135 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI12A6 | |
| Unconjugated | |
| IFIH1 | |
| 9130009C22Rik; AGS7; CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; helicard; Helicase with 2 CARD domains; Hlcd; IDDM19; IFIH1; interferon induced with helicase C domain 1; interferon induced with helicase C domain protein 1; interferon induced, with helicase C domain 1; interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA5; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RGD1560496; RH116; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116; SGMRT1 | |
| Full length human recombinant protein of human IFIH1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction