Learn More
IFIT1 Antibody (OTI3G8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71005C
Product Code. 18193802
  • € 795.00 / 0.10mL
Description
IFIT1 Monoclonal specifically detects IFIT1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFIT1
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
IFIT1
Full length recombinant protein of human IFIT1 (NP_000899) produced in HEK293T cells.
0.1 mL
3434.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3G8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
G10P1C56, GARG-16, IFI56IFI-56, IFI-56K, IFIT-1, IFNAI1ISG56, Interferon, alpha-inducible protein (MW 56kD), Interferon-induced 56 kDa protein, interferon-induced protein 56, interferon-induced protein with tetratricopeptide repeats 1, p56, RNM561
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Monkey