Learn More
IFIT2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98193APC
Product Code. 18633652
  • € 430.00 / 0.10mL
Description
IFIT2 Polyclonal antibody specifically detects IFIT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APC
PBS
3433
Immunohistochemistry-Paraffin
Primary
Store at 4C in the dark.