missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT2 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98193V
This item is not returnable.
View return policy
Description
IFIT2 Polyclonal antibody specifically detects IFIT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IFIT2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| cig42, G10P2IFI-54K, GARG-39, IFI-54, IFI54IFIT-2, IFI-54K, Interferon, alpha-inducible protein (MW 54kD), Interferon-induced 54 kDa protein, interferon-induced protein 54, interferon-induced protein with tetratricopeptide repeats 2, ISG54, ISG-54 K, ISG-54K, P54 | |
| Produced in rabbits immunized with E. coli-derived Human IFIT2 fragment. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction | |
| 3433 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction