Learn More
IFIT3 Antibody (OTI1G1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71006AF350
Product Code. 18116433
  • € 807.00 / 0.10mL
Description
IFIT3 Monoclonal specifically detects IFIT3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFIT3
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
IFIT3
Full length recombinant protein of human IFIT3 (NP_001540) produced in HEK293T cells.
Immunology
3437
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1G1
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49
Mouse
0.1 mL