missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFIT3 Recombinant Rabbit Monoclonal Antibody (HL246)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550544
This item is not returnable.
View return policy
Description
IFIT3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
IFN-induced antiviral protein which acts as an inhibitor of cellular as well as viral processes, cell migration, proliferation, signaling, and viral replication. Enhances MAVS-mediated host antiviral responses by serving as an adapter bridging TBK1 to MAVS which leads to the activation of TBK1 and phosphorylation of IRF3 and phosphorylated IRF3 translocates into nucleus to promote antiviral gene transcription. Exhibits an antiproliferative activity via the up-regulation of cell cycle negative regulators CDKN1A/p21 and CDKN1B/p27. Normally, CDKN1B/p27 turnover is regulated by COPS5, which binds CDKN1B/p27 in the nucleus and exports it to the cytoplasm for ubiquitin-dependent degradation. IFIT3 sequesters COPS5 in the cytoplasm, thereby increasing nuclear CDKN1B/p27 protein levels. Upregulates CDKN1A/p21 by downregulating MYC, a repressor of CDKN1A/p21. Can negatively regulate the apoptotic effects of IFIT2.
Specifications
| IFIT3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O14879 | |
| IFIT3 | |
| Recombinant fragment of human IFIT3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL246 | |
| Unconjugated | |
| IFIT3 | |
| cig41; CIG49; CIG-49; Garg49; GARG-49; Glucocorticoid-attenuated response gene 49 protein; Ifi49; IFI60; IFI-60K; IFIT3; IFIT-3; IFIT4; IFIT-4; interferon induced protein with tetratricopeptide repeats 3; interferon-induced 60 kDa protein; Interferon-induced protein with tetratricopeptide repeats 3; interferon-induced protein with tetratricopeptide repeats 4; interferon-stimulated protein 60; IRG2; Isg49; ISG60; ISG-60; P49; P60; retinoic acid-induced gene G protein; RIG-G; RP11-149I23.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3437 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction