missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM1 Antibody (001), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89229PCP
This item is not returnable.
View return policy
Description
IFITM1 Monoclonal antibody specifically detects IFITM1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IFITM1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 001 | |
| Immunohistochemistry-Paraffin | |
| CD225 antigen, CD225Leu-13 antigen, IFI17LEU13,9-27, interferon induced transmembrane protein 1 (9-27), Interferon-induced protein 17, interferon-induced transmembrane protein 1, Interferon-inducible protein 9-27 | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human IFITM1. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8519 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction