missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFITM2 Monoclonal Antibody (OTI2B3)
GREENER_CHOICE

Product Code. 15927094
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15927094 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15927094 Supplier Invitrogen™ Supplier No. MA527502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFITM2 Monoclonal Antibody for Western Blot, IHC (P)

IFN-induced antiviral protein which inhibits the entry of viruses to the host cell cytoplasm, permitting endocytosis, but preventing subsequent viral fusion and release of viral contents into the cytosol. Active against multiple viruses, including influenza A virus, SARS coronavirus (SARS-CoV), Marburg virus (MARV), Ebola virus (EBOV), Dengue virus (DNV), West Nile virus (WNV), human immunodeficiency virus type 1 (HIV-1) and vesicular stomatitis virus (VSV). Can inhibit: influenza virus hemagglutinin protein-mediated viral entry, MARV and EBOV GP1,2-mediated viral entry, SARS-CoV S protein-mediated viral entry and VSV G protein-mediated viral entry. Induces cell cycle arrest and mediates apoptosis by caspase activation and in p53-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFITM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFITM2
Gene Accession No. Q01629
Gene Alias 1-8D; dispanin subfamily A member 2c; DSPA2c; IFITM2; ifm2; interferon induced transmembrane protein 2; interferon-induced transmembrane protein 2; Interferon-inducible protein 1-8D
Gene Symbols IFITM2
Host Species Mouse
Immunogen Full length human recombinant protein of IFITM2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10581
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.