missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 1 Antibody (2-48), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08231AF594
This item is not returnable.
View return policy
Description
IFN-alpha 1 Monoclonal antibody specifically detects IFN-alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| IFN-alpha 1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| IFL;IFN;IFNA@;IFNA13;IFN-ALPHA;IFN-alphaD;Interferon alpha-1/13;leIF D | |
| Purified recombinant human IFN-alpha 1 (Uniprot: P01562) | |
| 100 μg | |
| Cancer, Immunology, Innate Immunity | |
| 3439 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 1948-02-01 00:00:00 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction