Learn More
IFN-alpha 1 Antibody (Chinese CDC patent anti-Interferon Alpha), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28179CL1
Product Code. 30153550
  • € 869.00 / 0.10mL
Description
IFN-alpha 1 Monoclonal antibody specifically detects IFN-alpha 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-alpha 1
Monoclonal
CoraFluor 1
IFL;IFN;IFNA@;IFNA13;IFN-ALPHA;IFN-alphaD;Interferon alpha-1/13;leIF D
IFNa1
0.1 mL
Cancer, Immunology, Innate Immunity
3439
Store at 4°C in the dark. Do not freeze.
IgG1
Flow Cytometry, ELISA, Functional Assay
Chinese CDC patent anti-Interferon Alpha
PBS
Human
Protein A purified