missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 2 Antibody (ST29), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2479AF532
This item is not returnable.
View return policy
Description
IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Human samples. It is validated for ELISA
Specifications
| IFN-alpha 2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
| Human interferon (HuIFN) produced by Namalwa cells (HuIFN-aN) | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cytokine Research | |
| 3440 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| ST29 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction