missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 4 Antibody (053), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89433AF405
This item is not returnable.
View return policy
Description
IFN-alpha 4 Monoclonal antibody specifically detects IFN-alpha 4 in Human samples. It is validated for ELISA
Specifications
| IFN-alpha 4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 053 | |
| ELISA | |
| IFN-alpha-4, IFN-alpha4a, INFA4, interferon alpha-4, Interferon alpha-4B, Interferon alpha-76, Interferon alpha-M1, interferon, alpha 4, MGC142200 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha 4 (NP_066546.1; Met1-Asp189). | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity | |
| 3441 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction