Learn More
IFN-alpha 4 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99055PE
Product Code. 18696795
  • € 407.00 / 0.10mL
Description
IFN-alpha 4 Polyclonal antibody specifically detects IFN-alpha 4 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA
IFN-alpha-4, IFN-alpha4a, INFA4, interferon alpha-4, Interferon alpha-4B, Interferon alpha-76, Interferon alpha-M1, interferon, alpha 4, MGC142200
3441
Store at 4C in the dark.