missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha B2/IFN-alpha 8 Antibody (003), DyLight 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89440UV
This item is not returnable.
View return policy
Description
IFN-alpha B2/IFN-alpha 8 Monoclonal antibody specifically detects IFN-alpha B2/IFN-alpha 8 in Human samples. It is validated for ELISA
Specifications
| IFN-alpha B2/IFN-alpha 8 | |
| Monoclonal | |
| DyLight 350 | |
| IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B', Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha B2/IFN-alpha 8 (NP_002161.2; Cys24-Glu189). | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity | |
| 3445 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 003 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction