missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha/beta R1 Antibody (MM0336-8H64), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11731B
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Monoclonal specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot.
Specifications
| IFN-alpha/beta R1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| IFNAR1 | |
| Human recombinant IFN-alpha/beta R1 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| MM0336-8H64 | |
| Western Blot, Flow Cytometry | |
| alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
| Mouse | |
| Protein G purified | |
| Neuroscience, Phospho Specific | |
| 3454 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction