missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha/beta R2 Antibody (122), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89447V
This item is not returnable.
View return policy
Description
IFN-alpha/beta R2 Monoclonal specifically detects IFN-alpha/beta R2 in Human samples. It is validated for Flow Cytometry.
Specifications
| IFN-alpha/beta R2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 122 | |
| Flow Cytometry | |
| beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFNARB, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha/beta R2 (NP_997468.1; Met1-Lys243). | |
| 0.1 mL | |
| Cancer | |
| 3455 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction