missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha/beta R2 Antibody (RM0251-5L16), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11743F
This item is not returnable.
View return policy
Description
IFN-alpha/beta R2 Monoclonal specifically detects IFN-alpha/beta R2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IFN-alpha/beta R2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| IFNAR2 | |
| Mouse recombinant IFN-alpha/beta R2 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0251-5L16 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFNARB, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
| Rat | |
| Protein A or G purified | |
| Cancer | |
| 3455 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction