missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha G/IFNA5 Antibody (003), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90891AF647
This item is not returnable.
View return policy
Description
IFN-alpha G/IFNA5 Monoclonal specifically detects IFN-alpha G/IFNA5 in Rat samples. It is validated for ELISA.
Specifications
| IFN-alpha G/IFNA5 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| IFN-alpha-5, IFN-alphaG, INFA5, interferon alpha-5, Interferon alpha-61, Interferon alpha-G, interferon, alpha 5, LeIF G | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Rat IFN-alpha G/IFNA5 (XP_001076062.1; Met1-Glu189). | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| ELISA | |
| 3 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3442 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction