missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IFN-alpha G/IFNA5 Antibody (08) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30740UV
This item is not returnable.
View return policy
Description
IFN-alpha G/IFNA5 Monoclonal antibody specifically detects IFN-alpha G/IFNA5 in Human samples. It is validated for ELISA
Specifications
| IFN-alpha G/IFNA5 | |
| Monoclonal | |
| DyLight 350 | |
| IFN-alpha-5, IFN-alphaG, INFA5, interferon alpha-5, Interferon alpha-61, Interferon alpha-G, interferon, alpha 5, LeIF G | |
| Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFN-alpha G/IFNA5 (Uniprot#: P01569; Met1-Glu189) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 08 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3442 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction