missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha G/IFNA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99235-50ul
This item is not returnable.
View return policy
Description
IFN-alpha G/IFNA5 Polyclonal antibody specifically detects IFN-alpha G/IFNA5 in Human samples. It is validated for ELISA
Specifications
| IFN-alpha G/IFNA5 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| IFN-alpha-5, IFN-alphaG, INFA5, interferon alpha-5, Interferon alpha-61, Interferon alpha-G, interferon, alpha 5, LeIF G | |
| Produced in rabbits immunized with purified, recombinant Human IFN-alpha G/IFNA5 (Uniprot#: P01569; Met1-Glu189) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3442 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction