missing translation for 'onlineSavingsMsg'
Learn More

IFN alpha-IFNAR-IN-1 hydrochloride, MedChemExpress

Product Code. 16431577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431577

Brand: Medchem Express HY12836A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IFN alpha-IFNAR-IN-1 hydrochloride is a nonpeptidic, low-molecular-weight inhibitor of the interaction between IFN-α and IFNAR; inhibit MVA-induced IFN-α responses by BM-pDCs (IC50=2-8 uM).

IFN alpha-IFNAR-IN-1 hydrochloride is a nonpeptidic, low-molecular-weight inhibitor of the interaction between IFN-α and IFNAR

Specifications

Chemical Name or Material IFN alpha-IFNAR-IN-1 hydrochloride
Physical Form Powder
CAS 2070014-98-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H18ClNS
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (316.60 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (52.78 mM; Need ultrasonic)
SMILES CNCC1=CC=CC=C1SC2=C(C=CC=C3)C3=CC=C2.[H]Cl
Molecular Weight (g/mol) 315.86
Formula Weight 315.86
Percent Purity 96.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.