Learn More
IFN-gamma Antibody (25723), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2851N
Product Code. 30060298
  • € 664.00 / 0.10mL
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-gamma
Monoclonal
Alexa Fluor 700
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF
25723
Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
E. coli-derived recombinant human IFN-gamma , Gln24-Gln166, Accession # AAP20098.1
0.1 mL
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
3458
Store at 4C in the dark.
IgG2b