Learn More
IFN-gamma Antibody (37895), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB485MFV450
Product Code. 18538796
  • € 695.00 / 0.10mL
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-gamma
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Rat
0.1 mL
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
3458
Store at 4C in the dark.
Western Blot, Immunohistochemistry, Neutralization, Flow Cytometry, CyTOF
37895
Western Blot, Immunohistochemistry, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
E. coli-derived recombinant mouse IFN-gamma