missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma R1/CD119 Antibody (RM0093-7K23), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22408AF350
This item is not returnable.
View return policy
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IFN-gamma R1/CD119 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IFNGR1 | |
| Mouse recombinant IFN-gamma R1/CD119 | |
| 0.1 mL | |
| 3459 | |
| Mouse | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0093-7K23 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of INFGR1. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction