missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma R2 Antibody (MOB47), PE/Cy7, Novus Biologicals™
Hamster Monoclonal Antibody
Brand: Novus Biologicals FAB773PECY7
This item is not returnable.
View return policy
Description
IFN-gamma R2 Monoclonal antibody specifically detects IFN-gamma R2 in Mouse samples. It is validated for Flow Cytometry
Specifications
| IFN-gamma R2 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3460 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| MOB47 | |
| Flow Cytometry | |
| AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
| Recombinant mouse IFN-gamma R2, Extracellular domain | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction