missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN gamma Rat, FITC, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
€ 131.25 - € 262.50
Specifications
| Antigen | IFN gamma |
|---|---|
| Clone | XMG1.2 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15902558
|
EnquireBio
QAB83-F-25ug |
25 μg |
€ 131.25
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15992548
|
EnquireBio
QAB83-F-100ug |
100 μg |
€ 262.50
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| IFN gamma | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| 15978 | |
| IgG1 κ | |
| Protein A/G Purified |
| XMG1.2 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| NP_032363 | |
| Ifng | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title