missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN gamma Rat, PE-Cy7, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
€ 162.00 - € 351.00
Specifications
Antigen | IFN gamma |
---|---|
Clone | XMG1.2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942558
|
EnquireBio
QAB83-PE7-25UG |
25 μg |
€ 162.00
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932558
|
EnquireBio
QAB83-PE7-100UG |
100 μg |
€ 351.00
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IFN gamma | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
15978 | |
IgG1 κ | |
Protein A/G Purified |
XMG1.2 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_032363 | |
Ifng | |
Primary | |
Store at 2-4°C. |