missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFN gamma Recombinant Rabbit Monoclonal Antibody (003), PerCP, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 131.00 - € 412.00
Specifications
| Antigen | IFN gamma |
|---|---|
| Clone | 003 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019976
|
Invitrogen™
MA552307 |
25 Tests |
€ 131.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019656
|
Invitrogen™
MA552306 |
100 Tests |
€ 412.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN gamma Recombinant Monoclonal antibody specifically detects IFN gamma in Human samples. It is validated for Flow CytometrySpecifications
| IFN gamma | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.09% sodium azide | |
| BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g | |
| Recombinant Human Interferon Gamma/IFN gamma/IFNG Protein (Gln 24-Gln166). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IFNG |
| 003 | |
| Flow Cytometry | |
| PerCP | |
| Rabbit | |
| Human | |
| P01579 | |
| 3458 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title