missing translation for 'onlineSavingsMsg'
Learn More

IFN-γ Mouse anti-Human, PE, Clone: 4S.B3, BD

Product Code. 15840729
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15840729 25 μg Each
15807778 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15840729 Supplier BD Biosciences Supplier No. 561056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 4S.B3 monoclonal antibody specifically binds to interferon-γ(IFN-γ). The immunogen used to generate this hybridoma was partially purified human IFN-γobtained from supernatants of human PBMC stimulated with Staphylococcus aureus . Interferon-γ(IFN-γ) is a potent multifunctional cytokine that is produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ + T cells. IFN- γexerts its biological effects through specific binding to the high-affinity IFN-γReceptor Complex comprised of IFN-γRα (CD119) and ese influences can involve IFN- γ’s capacity to boost MHC class I and II expression by antigen-presenting cells as well as to direct effects on B cells and T cells themselves. Human IFN-γis a 14- 18 kDa glycoprotein containing 143 amino acid residues.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry
Classification Monoclonal
Clone 4S.B3
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IFN-γ from supernatants of S. aureus-stimulated PBMC
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.