missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-omega/IFNW1 Antibody (04), Novus Biologicals™
Mouse Monoclonal Antibody
€ 231.00 - € 307.65
Specifications
| Antigen | IFN-omega/IFNW1 |
|---|---|
| Clone | 04 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656856
|
Novus Biologicals
NBP3-06154-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677398
|
Novus Biologicals
NBP3-06154-50ul |
50 μg |
€ 231.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-omega/IFNW1 Monoclonal antibody specifically detects IFN-omega/IFNW1 in Human samples. It is validated for ELISASpecifications
| IFN-omega/IFNW1 | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFN-omega/IFNW1 (Uniprot#: P05000; Met1-Ser195). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 04 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 3467 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title