Learn More
Invitrogen™ IFNA14 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA597859
Product Code. 16590682
  • € 464.00 / 100µg
Description
IFNA14 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFNA14
Polyclonal
Unconjugated
IFNA14
IFNA14; IFN-alpha-14; IFN-alphaH; Interferon; interferon alpha 14; interferon alpha-14; Interferon alpha-H; interferon lambda-2-H; interferon, alpha 14; leIF H; LEIF2H; OTTMUSG00000007662
Rabbit
Protein G
RUO
3448, 404549
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
4 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
P01570
IFNA14
Recombinant Human Interferon alpha-14 protein (24-189AA).