Learn More
Invitrogen™ IFNA14 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103422
Product Code. 16639874
  • € 502.00 / 100µL
Description
Antibody detects endogenous levels of total IFN14.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFNA14
Polyclonal
Unconjugated
IFNA14
IFNA14; IFN-alpha-14; IFN-alphaH; Interferon; interferon alpha 14; interferon alpha-14; Interferon alpha-H; interferon lambda-2-H; interferon, alpha 14; leIF H; LEIF2H; OTTMUSG00000007662
Rabbit
Affinity chromatography
RUO
3448
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P01570
IFNA14
A synthesized peptide derived from human IFNA14(Accession P01570), corresponding to amino acid residues L105-M155.