missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNA2 Monoclonal Antibody (OTI12C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811910S
This item is not returnable.
View return policy
Description
IFNA2 Monoclonal antibody specifically detects IFNA2 in Human samples. It is validated for Western BlotSpecifications
| IFNA2 | |
| Monoclonal | |
| 0.5 to 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H257 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64170 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI12C7 | |
| Unconjugated | |
| CARD9 | |
| alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2 | |
| Full length human recombinant protein of human CARD9 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction