missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNA2 Monoclonal Antibody (OTI12C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811910
This item is not returnable.
View return policy
Description
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.Specifications
| IFNA2 | |
| Monoclonal | |
| Unconjugated | |
| IFNA2 | |
| IFN-alphaA, IFNA, IFNA2B, INFA2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3440 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI12C7 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01563 | |
| IFNA2 | |
| Human recombit protein fragment corresponding to amino acids 24-188 of human IFNA2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction