missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNAG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144128
This item is not returnable.
View return policy
Description
IFNAG Polyclonal Antibody for Western Blot, ELISA
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| IFNAG | |
| Polyclonal | |
| Unconjugated | |
| IFNAG | |
| IFNA7; Interferon | |
| Rabbit | |
| Protein G | |
| RUO | |
| 100329206 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P49877 | |
| IFNAG | |
| Recombinant Bovine Interferon alpha-G protein (24-189AA). | |
| 100 μg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction