missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNGR1 Monoclonal Antibody (9B1G4D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530665
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IFNGR1/CD119.
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.
Specifications
| IFNGR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P15260 | |
| IFNGR1 | |
| Recombinant Human IFNGR1/CD119 protein (Met1-Gly245). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 9B1G4D1 | |
| Unconjugated | |
| IFNGR1 | |
| antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar | |
| Mouse | |
| Protein A | |
| RUO | |
| 3459 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction