missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNGR1 Recombinant Rabbit Monoclonal Antibody (2B9Q1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535147
This item is not returnable.
View return policy
Description
IFNGR1 Recombinant Monoclonal Antibody for ELISA, ICC/IF, Western Blot
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.
Specifications
| IFNGR1 | |
| Recombinant Monoclonal | |
| 1.09 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 | |
| P15260, P15261 | |
| IFNGR1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 2B9Q1 | |
| Unconjugated | |
| IFNGR1 | |
| antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116465, 15979, 3459 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction