Learn More
IFNGR2 Mouse anti-Human, Clone: OTI1C2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506734
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal domit or X-linked inheritance.Specifications
| IFNGR2 | |
| Monoclonal | |
| Unconjugated | |
| IFNGR2 | |
| AF-1, IFGR2, IFNGT1, IMD28 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3460 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P38484 | |
| IFNGR2 | |
| Full length human recombit protein of human IFNGR2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.