missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFT122 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559742
This item is not returnable.
View return policy
Description
Immunogen sequence: ITKQADWARN IKEPKAAVEM YISAGEHVKA IEICGDHGWV DMLIDIARKL DKAEREPLLL CATYLKKLDS PGYAAETYLK MGDLK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| IFT122 | |
| Polyclonal | |
| Unconjugated | |
| IFT122 | |
| C86139; CED; CED1; hypothetical protein zgc:77217; Ift122; intraflagellar transport 122; intraflagellar transport 122 homolog; intraflagellar transport 122 homolog (Chlamydomonas); intraflagellar transport protein 122 homolog; sopb; SPG; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140; WDR10; WDR10p; WDR140; zgc:77217 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55764 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBG6 | |
| IFT122 | |
| Recombinant protein corresponding to Human IFT122. Recombinant protein control fragment (Product #RP-96421). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction